siRNA Details
| Gene Symbol: Olfr911-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146873.2 | NP_667084.2 | 1 | 61 | ||
| GenBank mRNA | BC127963.1 | - | 1 | 61 | 40134907 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr911-ps1| Gene Symbol: Olfr911-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146873.2 | NP_667084.2 | 1 | 61 | ||
| GenBank mRNA | BC127963.1 | - | 1 | 61 | 40134907 | |