siRNA Details
| Gene Symbol: Olfr1297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146888.1 | NP_667099.1 | 1 | 338 | ||
| GenBank mRNA | BC104072.2 | - | 1 | 338 | 40041728 | |
| BC104073.2 | - | 1 | 338 | 40041729 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1297| Gene Symbol: Olfr1297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146888.1 | NP_667099.1 | 1 | 338 | ||
| GenBank mRNA | BC104072.2 | - | 1 | 338 | 40041728 | |
| BC104073.2 | - | 1 | 338 | 40041729 | ||