siRNA Details
| Gene Symbol: Olfr1280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146908.1 | NP_667119.1 | 1 | 676 | ||
| GenBank mRNA | BC119051.1 | - | 1 | 719 | 8733800 | |
| BC119053.1 | - | 1 | 719 | 8733802 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1280| Gene Symbol: Olfr1280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146908.1 | NP_667119.1 | 1 | 676 | ||
| GenBank mRNA | BC119051.1 | - | 1 | 719 | 8733800 | |
| BC119053.1 | - | 1 | 719 | 8733802 | ||