siRNA Details
| Gene Symbol: Olfr1494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146990.1 | NP_667201.1 | 1 | 13 | ||
| GenBank mRNA | BC104099.1 | - | 1 | 13 | 40043096 | |
| BC104100.1 | - | 1 | 13 | 40043097 | ||
| BC106804.1 | - | 1 | 13 | 40042568 | ||

Olfr1494
Genomic Map