siRNA Details
| Gene Symbol: Olfr1054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147019.1 | NP_667230.1 | 1 | 771 | ||
| Gene Symbol: Olfr1085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146590.2 | NP_666801.2 | 1 | 774 | ||

Olfr1054
Genomic Map