siRNA Details
| Gene Symbol: Olfr283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147036.1 | NP_667247.1 | 1 | 471 | ||
| GenBank mRNA | BC125358.1 | - | 1 | 560 | 40129873 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr283| Gene Symbol: Olfr283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147036.1 | NP_667247.1 | 1 | 471 | ||
| GenBank mRNA | BC125358.1 | - | 1 | 560 | 40129873 | |