siRNA Details
| Gene Symbol: Olfr669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147043.1 | NP_667254.1 | 1 | 554 | ||
| GenBank mRNA | BC104300.2 | - | 1 | 554 | 40043424 | |
| BC104301.2 | - | 1 | 552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr669| Gene Symbol: Olfr669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147043.1 | NP_667254.1 | 1 | 554 | ||
| GenBank mRNA | BC104300.2 | - | 1 | 554 | 40043424 | |
| BC104301.2 | - | 1 | 552 | |||