siRNA Details
| Gene Symbol: Olfr679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147044.1 | NP_667255.1 | 1 | 603 | ||
| GenBank mRNA | BC127993.1 | - | 1 | 607 | 40135720 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr679| Gene Symbol: Olfr679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147044.1 | NP_667255.1 | 1 | 603 | ||
| GenBank mRNA | BC127993.1 | - | 1 | 607 | 40135720 | |