siRNA Details
Gene Symbol: Olfr600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147046.2 | NP_667257.2 | 1 | 77 | ||
GenBank mRNA | BC130247.1 | - | 1 | 81 | 40142365 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147046.2 | NP_667257.2 | 1 | 77 | ||
GenBank mRNA | BC130247.1 | - | 1 | 81 | 40142365 |