siRNA Details
| Gene Symbol: Olfr1352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147071.2 | NP_667282.1 | 2 | 634 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1352| Gene Symbol: Olfr1352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147071.2 | NP_667282.1 | 2 | 634 | ||