siRNA Details
| Gene Symbol: Olfr656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147075.1 | NP_667286.1 | 1 | 930 | ||
| GenBank mRNA | BC110472.1 | - | 1 | 930 | 40044290 | |
| BC110473.2 | - | 1 | 929 | 40044291 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr656| Gene Symbol: Olfr656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147075.1 | NP_667286.1 | 1 | 930 | ||
| GenBank mRNA | BC110472.1 | - | 1 | 930 | 40044290 | |
| BC110473.2 | - | 1 | 929 | 40044291 | ||