siRNA Details
| Gene Symbol: Olfr639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147084.1 | NP_667295.1 | 1 | 247 | ||
| GenBank mRNA | BC145837.1 | - | 1 | 297 | 40131140 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr639| Gene Symbol: Olfr639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147084.1 | NP_667295.1 | 1 | 247 | ||
| GenBank mRNA | BC145837.1 | - | 1 | 297 | 40131140 | |