siRNA Details
| Gene Symbol: Olfr974 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147107.1 | NP_667318.1 | 1 | 93 | ||
| GenBank mRNA | BC131914.1 | - | 1 | 140 | 40130191 | |
| BC132593.1 | - | 1 | 140 | 40130870 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr974| Gene Symbol: Olfr974 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147107.1 | NP_667318.1 | 1 | 93 | ||
| GenBank mRNA | BC131914.1 | - | 1 | 140 | 40130191 | |
| BC132593.1 | - | 1 | 140 | 40130870 | ||