siRNA Details
| Gene Symbol: Olfr586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147111.1 | NP_667322.1 | 1 | 83 | ||
| GenBank mRNA | BC101949.1 | - | 1 | 82 | 40043610 | |
| BC103663.1 | - | 1 | 82 | 40043468 | ||
| BC104726.1 | - | 1 | 82 | 40043611 | ||

Olfr586
Genomic Map