siRNA Details
| Gene Symbol: Abcg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011920.3 | NP_036050.1 | 9 | 1598 | ||
| GenBank mRNA | AF140218.1 | - | 8 | 1161 | ||
| BC053730.1 | - | 9 | 1610 | 30064535 | ||
| KF662864.1 | - | 9 | 1195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcg2| Gene Symbol: Abcg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011920.3 | NP_036050.1 | 9 | 1598 | ||
| GenBank mRNA | AF140218.1 | - | 8 | 1161 | ||
| BC053730.1 | - | 9 | 1610 | 30064535 | ||
| KF662864.1 | - | 9 | 1195 | |||