siRNA Details
| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190974.1 | NP_001177903.1 | 14 | 1941 | ||
| NM_001190975.1 | NP_001177904.1 | 13 | 1677 | |||
| NM_009465.4 | NP_033491.2 | 15 | 1968 | |||
| GenBank mRNA | AK033237.1 | - | 14 | 1910 | ||
| AK155567.1 | - | 16 | 1854 | |||
| AK161294.1 | - | 8 | 641 | |||
| AK170010.1 | - | 14 | 1915 | |||
| AK170897.1 | - | 14 | 1575 | |||
| AK200128.1 | - | 3 | 148 | |||
| BC008551.1 | - | 10 | 1058 | |||
| BC046618.1 | - | 15 | 1805 | 6494383 | ||
| BC050914.1 | - | 15 | 1776 | 6418252 | ||
| BC058230.1 | - | 15 | 1829 | 6313662 | ||
| X63535.1 | - | 15 | 1717 | |||

Axl
Genomic Map