siRNA Details
| Gene Symbol: Ceacam9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011927.4 | NP_036057.1 | 2 | 273 | ||
| GenBank mRNA | AK049543.1 | - | 2 | 245 | ||
| L49180.1 | - | 2 | 273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam9| Gene Symbol: Ceacam9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011927.4 | NP_036057.1 | 2 | 273 | ||
| GenBank mRNA | AK049543.1 | - | 2 | 245 | ||
| L49180.1 | - | 2 | 273 | |||