siRNA Details
| Gene Symbol: Ciao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025296.4 | NP_079572.2 | 5 | 712 | ||
| GenBank mRNA | AK002314.1 | - | 5 | 657 | ||
| AK004129.1 | - | 5 | 638 | |||
| AK028376.1 | - | 5 | 639 | |||
| BC004089.1 | - | 5 | 633 | 3593736 | ||

Ciao1
Genomic Map