siRNA Details
| Gene Symbol: Clcn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011929.2 | NP_036059.1 | 9, 10 | 711 | ||
| GenBank mRNA | AK045787.1 | - | 4, 5 | 298 | ||
| AK145053.1 | - | 9, 10 | 756 | |||
| AK165362.1 | - | 9, 10 | 758 | |||
| AK220319.1 | - | 4, 5 | 368 | |||

Clcn6
Genomic Map