siRNA Details
| Gene Symbol: Decr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011933.2 | NP_036063.1 | 8 | 894 | ||
| GenBank mRNA | AF155575.1 | - | 8 | 894 | ||
| AK137212.1 | - | 8 | 825 | |||
| AK149270.1 | - | 8 | 1019 | |||
| AK162961.1 | - | 8 | 1030 | |||
| BC021865.1 | - | 8 | 866 | 5123899 | ||

Decr2
Genomic Map