siRNA Details
| Gene Symbol: Esrrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243792.1 | NP_001230721.1 | 3 | 689 | ||
| NM_011935.3 | NP_036065.1 | 2 | 627 | |||
| GenBank mRNA | AB221580.1 | - | 2 | 461 | ||
| AF117254.1 | - | 2 | 524 | |||
| AK052731.1 | - | 3 | 668 | |||
| BC082324.1 | - | 2 | 595 | 6850748 | ||

Esrrg
Genomic Map