siRNA Details
| Gene Symbol: Map2k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009157.4 | NP_033183.1 | 11 | 1173 | ||
| GenBank mRNA | AK036928.1 | - | 11 | 1177 | ||
| AK044534.1 | - | 11 | 1165 | |||
| AK044698.1 | - | 11 | 1165 | |||
| AK139777.1 | - | 12 | 1196 | |||
| BC029833.1 | - | 9 | 924 | 3962163 | ||
| U18310.1 | - | 11 | 1148 | |||

Map2k4
Genomic Map