siRNA Details
| Gene Symbol: Map3k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011945.2 | NP_036075.2 | 8, 9 | 1499 | ||
| GenBank mRNA | AB014614.1 | - | 8, 9 | 1476 | ||
| AF117340.1 | - | 8, 9 | 1499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k1| Gene Symbol: Map3k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011945.2 | NP_036075.2 | 8, 9 | 1499 | ||
| GenBank mRNA | AB014614.1 | - | 8, 9 | 1476 | ||
| AF117340.1 | - | 8, 9 | 1499 | |||