siRNA Details
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163643.1 | NP_001157115.1 | 6, 7 | 1242 | ||
NM_009582.4 | NP_033608.3 | 4, 5 | 1066 | |||
GenBank mRNA | AK029882.1 | - | 4, 5 | 875 | ||
AK034374.1 | - | 6, 7 | 1058 | |||
AK166435.1 | - | 6, 7 | 1162 | |||
BC047158.1 | - | 3, 4 | 860 | 6494216 | ||
BC057572.1 | - | 6, 7 | 1066 | 6401321 | ||
U09541.2 | - | 1, 2 | 169 | |||
U14636.1 | - | 4, 5 | 898 | |||
U23789.1 | - | 3, 4 | 856 |
