siRNA Details
| Gene Symbol: Map3k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011946.3 | NP_036076.2 | 10 | 1245 | ||
| GenBank mRNA | U43186.1 | - | 10 | 1143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k2| Gene Symbol: Map3k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011946.3 | NP_036076.2 | 10 | 1245 | ||
| GenBank mRNA | U43186.1 | - | 10 | 1143 | ||