siRNA Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009316.1 | NP_033342.1 | 7 | 876 | ||
| NM_172688.3 | NP_766276.1 | 7 | 876 | |||
| GenBank mRNA | AK046565.1 | - | 7 | 842 | ||
| AK053911.1 | - | 7 | 862 | |||
| AK137046.1 | - | 7 | 877 | |||
| AK159277.1 | - | 7 | 840 | |||
| BC006665.1 | - | 7 | 867 | 3499247 | ||
| D76446.1 | - | 7 | 876 | |||

Map3k7
Genomic Map