siRNA Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009316.1 | NP_033342.1 | 9, 10 | 1112 | ||
| NM_172688.3 | NP_766276.1 | 9, 10 | 1112 | |||
| GenBank mRNA | AK046565.1 | - | 9, 10 | 1078 | ||
| AK053911.1 | - | 9, 10 | 1098 | |||
| AK133642.1 | - | 3, 4 | 239 | |||
| AK137046.1 | - | 9, 10 | 1113 | |||
| AK159277.1 | - | 9, 10 | 1076 | |||
| CV559615.1 | - | 2, 3 | 168 | |||

Map3k7
Genomic Map