siRNA Details
| Gene Symbol: Map3k8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007746.2 | NP_031772.1 | 4 | 802 | ||
| GenBank mRNA | AK140334.1 | - | 4 | 803 | ||
| AK149624.1 | - | 4 | 803 | |||
| AK149846.1 | - | 4 | 803 | |||
| AK157123.1 | - | 4 | 769 | |||
| BC125286.1 | - | 3 | 703 | 40129801 | ||
| BC137922.1 | - | 3 | 703 | 8860941 | ||
| D13759.1 | - | 4 | 802 | |||

Map3k8
Genomic Map