siRNA Details
| Gene Symbol: Mapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011952.2 | NP_036082.1 | 6 | 915 | ||
| GenBank mRNA | AK155287.1 | - | 5 | 857 | ||
| BC013754.1 | - | 6 | 748 | |||
| BC029712.1 | - | 6 | 909 | 5041517 | ||
| Z14249.1 | - | 6 | 886 | |||

Mapk3
Genomic Map