siRNA Details
| Gene Symbol: Mapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011952.2 | NP_036082.1 | 2, 3 | 369 | ||
| GenBank mRNA | AK155287.1 | - | 1, 2 | 311 | ||
| BC013754.1 | - | 2, 3 | 202 | |||
| BC029712.1 | - | 2, 3 | 363 | 5041517 | ||
| BQ929318.1 | - | 2, 3 | 369 | |||
| Z14249.1 | - | 2, 3 | 340 | |||

Mapk3
Genomic Map