siRNA Details
| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016700.4 | NP_057909.1 | 3 | 407 | ||
| GenBank mRNA | AB219977.1 | - | 2 | 216 | ||
| AK030767.1 | - | 3 | 409 | |||
| AK032486.1 | - | 3 | 405 | |||
| AK043102.1 | - | 3 | 407 | |||
| AK085745.1 | - | 3 | 407 | |||
| AK139498.1 | - | 3 | 395 | |||
| BB619635.1 | - | 3 | 409 | |||
| BC053027.1 | - | 3 | 377 | 5694771 | ||

Mapk8
Genomic Map