siRNA Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011960.2 | NP_036090.2 | 3 | 1303 | ||
| GenBank mRNA | AF079557.1 | - | 3 | 1052 | ||
| AK036656.1 | - | 3 | 1326 | |||
| AK160742.1 | - | 2 | 777 | |||
| AK163388.1 | - | 3 | 1330 | |||
| AK164013.1 | - | 3 | 1290 | |||
| BC050892.1 | - | 3 | 1302 | |||
| BC053037.1 | - | 3 | 1286 | |||
| BC059827.1 | - | 3 | 1312 | |||

Parg
Genomic Map