siRNA Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011960.2 | NP_036090.2 | 17 | 2938 | ||
| GenBank mRNA | AF079557.1 | - | 17 | 2687 | ||
| AK034542.1 | - | 10 | 870 | |||
| AK036656.1 | - | 17 | 2961 | |||
| AK088708.1 | - | 14 | 1293 | |||
| AK139114.1 | - | 2 | 153 | |||
| AK164013.1 | - | 17 | 2926 | |||
| BC050892.1 | - | 17 | 2937 | |||
| BC053037.1 | - | 17 | 2922 | |||
| BC059827.1 | - | 17 | 2947 | |||
| EF382673.1 | - | 15 | 1647 | |||
| JQ890227.1 | - | 14 | 1444 | |||

Parg
Genomic Map