siRNA Details
| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011962.3 | NP_036092.1 | 10 | 1694 | ||
| GenBank mRNA | AF046783.1 | - | 10 | 1102 | ||
| AK013195.1 | - | 10 | 1240 | |||
| AK033360.1 | - | 10 | 1239 | |||
| AK053320.1 | - | 10 | 1336 | |||
| AK088948.1 | - | 10 | 1111 | |||
| AK157741.1 | - | 9 | 1344 | |||
| AK183180.1 | - | 2 | 45 | |||
| AK183516.1 | - | 2 | 45 | |||
| BC043047.1 | - | 10 | 1693 | 5703967 | ||
| BC054734.1 | - | 10 | 1694 | 5720686 | ||

Plod3
Genomic Map