siRNA Details
| Gene Symbol: Psma4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011966.3 | NP_036096.1 | 5, 6 | 351 | ||
| GenBank mRNA | AF060093.1 | - | 5, 6 | 336 | ||
| AK085937.1 | - | 5, 6 | 409 | |||
| AK150565.1 | - | 5, 6 | 396 | |||
| AK166618.1 | - | 5, 6 | 390 | |||
| AK168150.1 | - | 5, 6 | 367 | |||
| AK168838.1 | - | 5, 6 | 410 | |||
| BC001982.1 | - | 5, 6 | 351 | 3592024 | ||

Psma4
Genomic Map