siRNA Details
Gene Symbol: Psma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289476.1 | NP_001276405.1 | 5 | 1262 | ||
NM_011969.2 | NP_036099.1 | 5 | 623 | |||
GenBank mRNA | AF019662.1 | - | 5 | 561 | ||
AK088615.1 | - | 5 | 568 | |||
AK146765.1 | - | 5 | 580 | |||
AK165563.1 | - | 5 | 576 | |||
AK168234.1 | - | 5 | 594 | |||
BC008222.1 | - | 5 | 556 | 2581897 | ||
BC014752.1 | - | 5 | 1210 | |||
DV049235.1 | - | 5 | 592 |
