siRNA Details
| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011970.4 | NP_036100.3 | 3 | 348 | ||
| GenBank mRNA | AF060090.1 | - | 3 | 236 | ||
| AK078487.1 | - | 3 | 327 | |||
| BB853639.1 | - | 3 | 361 | |||
| BC008265.1 | - | 3 | 275 | 2811010 | ||
| CB321399.1 | - | 3 | 344 | |||

Psmb2
Genomic Map