siRNA Details
| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011970.4 | NP_036100.3 | 4 | 457 | ||
| GenBank mRNA | AF060090.1 | - | 4 | 345 | ||
| AK078487.1 | - | 4 | 436 | |||
| BC008265.1 | - | 4 | 384 | 2811010 | ||
| CB321399.1 | - | 4 | 453 | |||

Psmb2
Genomic Map