siRNA Details
| Gene Symbol: Mok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011973.2 | NP_036103.1 | 3 | 379 | ||
| GenBank mRNA | AB022695.1 | - | 3 | 379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mok| Gene Symbol: Mok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011973.2 | NP_036103.1 | 3 | 379 | ||
| GenBank mRNA | AB022695.1 | - | 3 | 379 | ||