siRNA Details
| Gene Symbol: Cul3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016716.4 | NP_057925.1 | 14 | 2083 | ||
| GenBank mRNA | AF129738.1 | - | 14 | 1973 | ||
| AK129174.1 | - | 15 | 2471 | |||
| BC027304.1 | - | 14 | 2108 | 3981170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cul3| Gene Symbol: Cul3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016716.4 | NP_057925.1 | 14 | 2083 | ||
| GenBank mRNA | AF129738.1 | - | 14 | 1973 | ||
| AK129174.1 | - | 15 | 2471 | |||
| BC027304.1 | - | 14 | 2108 | 3981170 | ||