siRNA Details
| Gene Symbol: Cul3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016716.4 | NP_057925.1 | 6, 7 | 1012 | ||
| GenBank mRNA | AF129738.1 | - | 6, 7 | 902 | ||
| AK129174.1 | - | 7, 8 | 1400 | |||
| BC027304.1 | - | 6, 7 | 1037 | 3981170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cul3| Gene Symbol: Cul3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016716.4 | NP_057925.1 | 6, 7 | 1012 | ||
| GenBank mRNA | AF129738.1 | - | 6, 7 | 902 | ||
| AK129174.1 | - | 7, 8 | 1400 | |||
| BC027304.1 | - | 6, 7 | 1037 | 3981170 | ||