siRNA Details
| Gene Symbol: Ror2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013846.3 | NP_038874.3 | 9 | 1696 | ||
| GenBank mRNA | AB010384.1 | - | 9 | 1464 | ||
| AK031112.1 | - | 9 | 1666 | |||
| AK031789.1 | - | 9 | 1453 | |||
| AK083538.1 | - | 9 | 1672 | |||
| AK084752.1 | - | 9 | 1662 | |||
| AK157705.1 | - | 9 | 1712 | |||
| BC030848.1 | - | 7 | 1037 | |||
| BC070436.1 | - | 9 | 1696 | 30535615 | ||

Ror2
Genomic Map