siRNA Details
| Gene Symbol: Irak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029926.5 | NP_084202.2 | 6, 7 | 891 | ||
| GenBank mRNA | AF445803.1 | - | 6, 7 | 879 | ||
| AK028837.1 | - | 6, 7 | 832 | |||
| AK029028.1 | - | 6, 7 | 892 | |||
| AK139992.1 | - | 880 | ||||
| BC051676.1 | - | 6, 7 | 884 | 30017484 | ||

Irak4
Genomic Map