siRNA Details
| Gene Symbol: Snx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153680.2 | NP_710147.1 | 5 | 455 | ||
| GenBank mRNA | AK047265.1 | - | 5 | 455 | ||
| AK077650.1 | - | 5 | 457 | |||
| AK077780.1 | - | 5 | 457 | |||
| AK156299.1 | - | 5 | 506 | |||
| AK219724.1 | - | 2 | 100 | |||
| BC023732.1 | - | 5 | 471 | 5352355 | ||
| BC026571.1 | - | 5 | 455 | 4986268 | ||
| BC029675.1 | - | 5 | 442 | |||

Snx17
Genomic Map