siRNA Details
| Gene Symbol: Scml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172938.3 | NP_766526.3 | 8 | 1562 | ||
| GenBank mRNA | AF397912.1 | - | 1 | 99 | ||
| AK034171.1 | - | 8 | 1401 | |||
| AK040597.1 | - | 3 | 1210 | |||
| AK043500.1 | - | 3 | 1093 | |||
| AK047972.1 | - | 6 | 988 | |||
| AK141826.1 | - | 3 | 586 | |||
| AK172160.1 | - | 4 | 625 | |||
| AK220309.1 | - | 6 | 946 | |||
| BC024570.1 | - | 3 | 518 | |||
| BC039567.1 | - | 1 | 85 | |||
| BC043310.1 | - | 8 | 1357 | 5067233 | ||

Scml4
Genomic Map