siRNA Details
Gene Symbol: Zkscan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130529.2 | NP_001124001.1 | 4 | 738 | ||
NM_001291014.1 | NP_001277943.1 | 4 | 785 | |||
NM_172941.4 | NP_766529.3 | 4 | 785 | |||
GenBank mRNA | AK051711.1 | - | 4 | 367 | ||
AK139346.1 | - | 4 | 1028 | |||
AK155196.1 | - | 4 | 315 | |||
AK155490.1 | - | 4 | 367 | |||
AK158448.1 | - | 4 | 322 | |||
AK170924.1 | - | 4 | 315 | |||
AK170989.1 | - | 4 | 320 | |||
AK171190.1 | - | 4 | 367 | |||
AK178512.1 | - | 2 | 138 | |||
AK182740.1 | - | 223 | ||||
AK190897.1 | - | 3 | 163 | |||
AK209254.1 | - | 3 | 163 | |||
AK218924.1 | - | 1 | 53 | |||
AY302242.1 | - | 3 | 266 | |||
BC040205.1 | - | 3 | 270 | 4947290 | ||
BY753203.1 | - | 4 | 319 | |||
CN530086.1 | - | 2 | 185 |
