siRNA Details
| Gene Symbol: Rapgefl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080925.1 | NP_001074394.1 | 13 | 2154 | ||
| GenBank mRNA | AK045359.1 | - | 1 | 75 | ||
| BC080279.1 | - | 12 | 1222 | |||
| BC130266.1 | - | 14 | 1430 | 40143028 | ||

Rapgefl1
Genomic Map