siRNA Details
| Gene Symbol: Rapgefl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080925.1 | NP_001074394.1 | 11 | 1995 | ||
| GenBank mRNA | BC080279.1 | - | 10 | 1063 | ||
| BC130266.1 | - | 12 | 1271 | 40143028 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rapgefl1| Gene Symbol: Rapgefl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080925.1 | NP_001074394.1 | 11 | 1995 | ||
| GenBank mRNA | BC080279.1 | - | 10 | 1063 | ||
| BC130266.1 | - | 12 | 1271 | 40143028 | ||