siRNA Details
| Gene Symbol: Krt222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172946.2 | NP_766534.1 | 1 | 57 | ||
| GenBank mRNA | AK031954.1 | - | 1 | 58 | ||
| AK044068.1 | - | 1 | 76 | |||
| AK138778.1 | - | 1 | 113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt222| Gene Symbol: Krt222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172946.2 | NP_766534.1 | 1 | 57 | ||
| GenBank mRNA | AK031954.1 | - | 1 | 58 | ||
| AK044068.1 | - | 1 | 76 | |||
| AK138778.1 | - | 1 | 113 | |||