siRNA Details
| Gene Symbol: Krt222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172946.2 | NP_766534.1 | 5 | 635 | ||
| GenBank mRNA | AK031954.1 | - | 5 | 636 | ||
| AK044068.1 | - | 5 | 735 | |||
| AK138778.1 | - | 5 | 691 | |||
| BC079890.1 | - | 5 | 629 | 30360088 | ||

Krt222
Genomic Map